Product Details

SNP ID
rs149158957
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:17664557 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAACAAATAATACAGGGAGCAATAA[A/G]ATTCATGTTTTCTAAAATAAAAAGT
Phenotype
MIM: 609387 MIM: 600817
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SMC6 PubMed Links

Gene Details

Gene
SMC6
Gene Name
structural maintenance of chromosomes 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142286.1 4342 UTR 3 NP_001135758.1
NM_024624.5 4342 UTR 3 NP_078900.1
XM_011533107.2 4342 UTR 3 XP_011531409.1
XM_011533108.2 4342 UTR 3 XP_011531410.1
XM_017004913.1 4342 UTR 3 XP_016860402.1
XM_017004914.1 4342 UTR 3 XP_016860403.1
XM_017004915.1 4342 UTR 3 XP_016860404.1
XM_017004916.1 4342 UTR 3 XP_016860405.1
Gene
VSNL1
Gene Name
visinin like 1
There are no transcripts associated with this gene.

View Full Product Details