Product Details

SNP ID
rs149306780
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:162144154 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGCGGCCAAGTTCTTCAACAATGG[C/T]GACCTCTTCTGGGAAACTAAGAAAA
Phenotype
MIM: 138030
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GCG PubMed Links

Gene Details

Gene
GCG
Gene Name
glucagon
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002054.4 665 Missense Mutation NP_002045.1
Gene
LOC101929532
Gene Name
uncharacterized LOC101929532
There are no transcripts associated with this gene.

View Full Product Details