Product Details

SNP ID
rs149652377
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:215319677 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTTTTAAATTAGGAATCCTAGCTC[A/G]TAATATTCCAGAAGATAATGCTGAC
Phenotype
MIM: 601731
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ATIC PubMed Links

Gene Details

Gene
ATIC
Gene Name
5-aminoimidazole-4-carboxamide ribonucleotide formyltransferase/IMP cyclohydrolase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004044.6 423 Missense Mutation CAT,CGT H79R NP_004035.2
XM_017004187.1 423 Missense Mutation CAT,CGT H79R XP_016859676.1

View Full Product Details