Product Details

SNP ID
rs149852767
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:28851263 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACATGAAAGAGCAAGTTCACAGGTG[C/T]GAATTCCATCTAAAAGTTCAATAAC
Phenotype
MIM: 614029
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SPDYA PubMed Links

Gene Details

Gene
SPDYA
Gene Name
speedy/RINGO cell cycle regulator family member A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001008779.1 859 Intron NP_001008779.1
NM_001142634.1 859 Intron NP_001136106.1
NM_182756.3 859 Intron NP_877433.2
Gene
TRMT61B
Gene Name
tRNA methyltransferase 61B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017910.3 859 Missense Mutation CAC,CGC H374R NP_060380.3
XM_005264393.2 859 Missense Mutation CAC,CGC H374R XP_005264450.1
XM_006712037.2 859 Missense Mutation CAC,CGC H276R XP_006712100.1
XM_017004401.1 859 Missense Mutation CAC,CGC H276R XP_016859890.1
XM_017004402.1 859 Intron XP_016859891.1
XM_017004403.1 859 Missense Mutation CAC,CGC H138R XP_016859892.1

View Full Product Details