Product Details

SNP ID
rs150264684
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:45389452 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAAAGTTTTGCTTCTGTTACATTT[C/T]GTATGGGAATCAGCCCAGATTTCCC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SRBD1 PubMed Links

Gene Details

Gene
SRBD1
Gene Name
S1 RNA binding domain 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018079.4 2476 Missense Mutation CAA,CGA Q949R NP_060549.4
XM_011532946.1 2476 Missense Mutation CAA,CGA Q933R XP_011531248.1
XM_017004417.1 2476 Intron XP_016859906.1
XM_017004418.1 2476 Missense Mutation CAA,CGA Q452R XP_016859907.1

View Full Product Details