Product Details

SNP ID
rs150432172
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:230908305 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGCTCTCTTCCTCACCCTTCCCTCC[A/G]TCCACCTAGGAGGAGGTGTACAGGG
Phenotype
MIM: 604107
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GPR55 PubMed Links

Gene Details

Gene
GPR55
Gene Name
G protein-coupled receptor 55
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005683.3 2851 UTR 3 NP_005674.2
XM_005246952.3 2851 UTR 3 XP_005247009.1
XM_011512175.2 2851 UTR 3 XP_011510477.1
XM_011512176.2 2851 UTR 3 XP_011510478.1
Gene
LOC151484
Gene Name
uncharacterized LOC151484
There are no transcripts associated with this gene.

View Full Product Details