Product Details
- SNP ID
-
rs150711518
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:134964910 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCTGGAGAGCAGAAGGTCGCTCATG[C/G]TGGTCCCTAAGAAGGGAAAAACACA
- Phenotype
-
MIM: 603862
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
CCNT2
PubMed Links
Gene Details
- Gene
- CCNT2
- Gene Name
- cyclin T2
There are no transcripts associated with this gene.
- Gene
- MAP3K19
- Gene Name
- mitogen-activated protein kinase kinase kinase 19
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001018044.2 |
3619 |
Missense Mutation |
CAC,CAG |
H1196Q |
NP_001018054.1 |
NM_001018046.2 |
3619 |
Missense Mutation |
CAC,CAG |
H491Q |
NP_001018056.1 |
NM_001018047.2 |
3619 |
Missense Mutation |
CAC,CAG |
H443Q |
NP_001018057.1 |
NM_001282883.1 |
3619 |
Missense Mutation |
CAC,CAG |
H441Q |
NP_001269812.1 |
NM_001321177.1 |
3619 |
Intron |
|
|
NP_001308106.1 |
NM_025052.4 |
3619 |
Missense Mutation |
CAC,CAG |
H1309Q |
NP_079328.3 |
XM_011511891.2 |
3619 |
Missense Mutation |
CAC,CAG |
H1326Q |
XP_011510193.1 |
XM_011511892.1 |
3619 |
Missense Mutation |
CAC,CAG |
H1309Q |
XP_011510194.1 |
XM_011511894.2 |
3619 |
Missense Mutation |
CAC,CAG |
H1281Q |
XP_011510196.1 |
XM_011511896.2 |
3619 |
Missense Mutation |
CAC,CAG |
H1214Q |
XP_011510198.1 |
XM_011511897.2 |
3619 |
Missense Mutation |
CAC,CAG |
H1214Q |
XP_011510199.1 |
XM_017005003.1 |
3619 |
Missense Mutation |
CAC,CAG |
H1309Q |
XP_016860492.1 |
XM_017005004.1 |
3619 |
Missense Mutation |
CAC,CAG |
H1281Q |
XP_016860493.1 |
XM_017005005.1 |
3619 |
Missense Mutation |
CAC,CAG |
H1281Q |
XP_016860494.1 |
XM_017005006.1 |
3619 |
Missense Mutation |
CAC,CAG |
H1187Q |
XP_016860495.1 |
XM_017005007.1 |
3619 |
Intron |
|
|
XP_016860496.1 |
View Full Product Details