Product Details

SNP ID
rs151070769
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:218360587 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGGCCCTCAGTTCCTCATCCTCCC[A/C]ATGGAACGGAAGATGAGTTTGCTGA
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CATIP PubMed Links

Gene Details

Gene
CATIP
Gene Name
ciliogenesis associated TTC17 interacting protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001320865.1 587 Silent Mutation CCA,CCC P141P NP_001307794.1
NM_198559.1 587 Silent Mutation CCA,CCC P130P NP_940961.1
XM_005246539.4 587 Silent Mutation CCA,CCC P99P XP_005246596.1
XM_005246541.4 587 Silent Mutation CCA,CCC P32P XP_005246598.1
XM_011511148.2 587 Silent Mutation CCA,CCC P141P XP_011509450.1
XM_011511149.2 587 Silent Mutation CCA,CCC P15P XP_011509451.1
XM_011511150.2 587 Silent Mutation CCA,CCC P15P XP_011509452.1
XM_017004053.1 587 UTR 5 XP_016859542.1
Gene
CATIP-AS1
Gene Name
CATIP antisense RNA 1
There are no transcripts associated with this gene.

Gene
CATIP-AS2
Gene Name
CATIP antisense RNA 2
There are no transcripts associated with this gene.

View Full Product Details