Product Details

SNP ID
rs151273922
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:218673945 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATCCTAGAAGATGACGGAAAACTT[C/T]CTGAAGACCAGGTATGCTTTCTGCC
Phenotype
MIM: 616014 MIM: 607652
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RNF25 PubMed Links

Gene Details

Gene
RNF25
Gene Name
ring finger protein 25
There are no transcripts associated with this gene.

Gene
STK36
Gene Name
serine/threonine kinase 36
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001243313.1 239 Missense Mutation CCT,TCT P98S NP_001230242.1
NM_015690.4 239 Missense Mutation CCT,TCT P98S NP_056505.2
XM_005246464.1 239 Missense Mutation CCT,TCT P98S XP_005246521.1
XM_011510959.1 239 Missense Mutation CCT,TCT P98S XP_011509261.1
XM_011510960.1 239 Missense Mutation CCT,TCT P44S XP_011509262.1
XM_011510961.1 239 Intron XP_011509263.1
XM_017003804.1 239 Missense Mutation CCT,TCT P98S XP_016859293.1
XM_017003805.1 239 UTR 5 XP_016859294.1

View Full Product Details