Product Details

SNP ID
rs151283384
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:183080734 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAGATTGGGGTTGGAAACCTCCCC[A/G]GGATAGTGGGAGAGTCCTGGGAGGA
Phenotype
MIM: 611437
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DUSP19 PubMed Links

Gene Details

Gene
DUSP19
Gene Name
dual specificity phosphatase 19
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142314.1 Intron NP_001135786.1
NM_001321519.1 Intron NP_001308448.1
NM_080876.3 Intron NP_543152.1

View Full Product Details