Product Details

SNP ID
rs138281794
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:129291285 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACCTTTTTAAAATGTGCAATTCAG[A/G]AATGTTTTAGTATGTTCAAAGTATG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HMCES PubMed Links

Gene Details

Gene
HMCES
Gene Name
5-hydroxymethylcytosine (hmC) binding, ES cell-specific
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001006109.1 Intron NP_001006109.1
NM_020187.2 Intron NP_064572.2
XM_005247636.3 Intron XP_005247693.1
XM_005247637.3 Intron XP_005247694.1
XM_017006877.1 Intron XP_016862366.1

View Full Product Details