Product Details
- SNP ID
-
rs138683352
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:186048758 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GTGTCCTCCTCGCTGAGGTGGCACT[C/T]GGACTCTGCCTTCAGGAACGGACGC
- Phenotype
-
MIM: 601600
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
ETV5
PubMed Links
Gene Details
- Gene
- ETV5
- Gene Name
- ETS variant 5
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_004454.2 |
1661 |
Missense Mutation |
AAG,GAG |
K472E |
NP_004445.1 |
View Full Product Details