Product Details

SNP ID
rs138703955
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:48469374 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGGGCACTCACCAGCCAGGGTCTC[A/G]TGGTAGGCCGGTGGGCCCATGGGCT
Phenotype
MIM: 606605 MIM: 607290 MIM: 606609
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ATRIP PubMed Links

Gene Details

Gene
ATRIP
Gene Name
ATR interacting protein
There are no transcripts associated with this gene.

Gene
SHISA5
Gene Name
shisa family member 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001272065.1 946 Intron NP_001258994.1
NM_001272066.1 946 Silent Mutation CAC,CAT H179H NP_001258995.1
NM_001272067.1 946 Silent Mutation CAC,CAT H179H NP_001258996.1
NM_001272068.1 946 Silent Mutation CAC,CAT H179H NP_001258997.1
NM_001272082.1 946 Silent Mutation CAC,CAT H107H NP_001259011.1
NM_001272083.1 946 Silent Mutation CAC,CAT H107H NP_001259012.1
NM_016479.4 946 Intron NP_057563.3
XM_005265205.1 946 Silent Mutation CAC,CAT H179H XP_005265262.1
XM_006713188.2 946 Silent Mutation CAC,CAT H179H XP_006713251.1
XM_011533796.1 946 Silent Mutation CAC,CAT H179H XP_011532098.1
Gene
TREX1
Gene Name
three prime repair exonuclease 1
There are no transcripts associated with this gene.

View Full Product Details