Product Details

SNP ID
rs138729991
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:194341194 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCCCTGCTGAGGAGAGAGCTGGAC[A/G]TTCAGCCAGCGACACTGGGCCTGGT
Phenotype
MIM: 603104
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CPN2 PubMed Links

Gene Details

Gene
CPN2
Gene Name
carboxypeptidase N subunit 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080513.3 1599 Silent Mutation AAC,AAT N503N NP_001073982.3
NM_001291988.1 1599 Silent Mutation AAC,AAT N503N NP_001278917.1
XM_005269280.4 1599 Silent Mutation AAC,AAT N503N XP_005269337.1

View Full Product Details