Product Details

SNP ID
rs139041816
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:49721805 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACTCGCCAATAGACTTGTGGGGCA[A/G]CACGCTGGCTCCGTTGAGGTAGAGC
Phenotype
MIM: 615691 MIM: 615320 MIM: 606991 MIM: 614472
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
AMIGO3 PubMed Links

Gene Details

Gene
AMIGO3
Gene Name
adhesion molecule with Ig like domain 3
There are no transcripts associated with this gene.

Gene
GMPPB
Gene Name
GDP-mannose pyrophosphorylase B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_013334.3 1278 Silent Mutation CTG,TTG L371L NP_037466.2
NM_021971.2 1278 Silent Mutation CTG,TTG L344L NP_068806.1
Gene
IP6K1
Gene Name
inositol hexakisphosphate kinase 1
There are no transcripts associated with this gene.

Gene
RNF123
Gene Name
ring finger protein 123
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022064.4 1278 Intron NP_071347.2
XM_006713285.1 1278 Intron XP_006713348.1
XM_011533995.1 1278 Intron XP_011532297.1
XM_017007018.1 1278 Intron XP_016862507.1

View Full Product Details