Product Details
- SNP ID
-
rs139369374
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:133805885 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGACTCGCGCCGGGTGGCAGATGGC[C/G]GCGGTGCCGGGGGCACCTTCCAGCC
- Phenotype
-
MIM: 616883
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
SRPRB
PubMed Links
Gene Details
- Gene
- SRPRB
- Gene Name
- SRP receptor beta subunit
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_021203.3 |
322 |
Missense Mutation |
CGC,GGC |
R13G |
NP_067026.3 |
View Full Product Details