Product Details
- SNP ID
-
rs139391516
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:186050091 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AATAAAATAATAACCCCTTCCTTAC[A/C]CTGGATGGCACATTATTATTTCTGA
- Phenotype
-
MIM: 601600
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
ETV5
PubMed Links
Gene Details
- Gene
- ETV5
- Gene Name
- ETS variant 5
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_004454.2 |
|
Intron |
|
|
NP_004445.1 |
View Full Product Details