Product Details
- SNP ID
-
rs139498139
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:71773651 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTCATACATTTGCACACAGATGCCA[A/G]GAGTCAAAATGCTGGGAAAAAACTT
- Phenotype
-
MIM: 607002
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
PROK2
PubMed Links
Gene Details
- Gene
- PROK2
- Gene Name
- prokineticin 2
View Full Product Details