Product Details

SNP ID
rs139608844
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:49001572 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATGAGCACTATGCAGGTCAGCCAG[C/T]TGGACCTCTTCCGGCTGCTGGACCA
Phenotype
MIM: 614584 MIM: 606031
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
P4HTM PubMed Links

Gene Details

Gene
P4HTM
Gene Name
prolyl 4-hydroxylase, transmembrane
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_177938.2 920 Silent Mutation CTG,TTG L191L NP_808807.2
NM_177939.2 920 Silent Mutation CTG,TTG L191L NP_808808.1
Gene
WDR6
Gene Name
WD repeat domain 6
There are no transcripts associated with this gene.

View Full Product Details