Product Details
- SNP ID
-
rs139751187
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:184710962 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCTCATACTGGCTTCAGCTCTGGCC[C/T]TGGCTCTGGCCCTGTCGGCCTCGTC
- Phenotype
-
MIM: 609267
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
MAGEF1
PubMed Links
Gene Details
- Gene
- MAGEF1
- Gene Name
- MAGE family member F1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_022149.4 |
1087 |
Missense Mutation |
AAG,AGG |
K287R |
NP_071432.2 |
View Full Product Details