Product Details

SNP ID
rs139947240
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:112991273 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATCTTCACGGCCACTGAACGGAAC[A/C]GCATCGACTACGTCAGCTCCGCCGT
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
GTPBP8 PubMed Links

Gene Details

Gene
GTPBP8
Gene Name
GTP binding protein 8 (putative)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014170.2 321 Missense Mutation AGC,CGC S92R NP_054889.2
NM_138485.1 321 Missense Mutation AGC,CGC S92R NP_612494.1

View Full Product Details