Product Details

SNP ID
rs140566252
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:38697850 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGATCTTATCTCCAGGGACCAAAGG[A/C]AGGTCCATCTGGATCAGTATATTTC
Phenotype
MIM: 604427
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
SCN10A PubMed Links

Gene Details

Gene
SCN10A
Gene Name
sodium voltage-gated channel alpha subunit 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001293306.2 5076 Silent Mutation CTG,CTT L1789L NP_001280235.2
NM_001293307.2 5076 Silent Mutation CTG,CTT L1692L NP_001280236.2
NM_006514.3 5076 Silent Mutation CTG,CTT L1790L NP_006505.3
XM_005265371.3 5076 Silent Mutation CTG,CTT L1793L XP_005265428.1
XM_011533993.2 5076 Silent Mutation CTG,CTT L1792L XP_011532295.1
XM_011533994.2 5076 Silent Mutation CTG,CTT L1695L XP_011532296.1

View Full Product Details