Product Details

SNP ID
rs140781331
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:190388148 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCTCCAGGACCCCACTGTTGGTTA[C/T]AGCCTGTTTGTATTATTCTTACTGC
Phenotype
MIM: 603959
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CLDN16 PubMed Links

Gene Details

Gene
CLDN16
Gene Name
claudin 16
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006580.3 277 Missense Mutation ACA,ATA T10I NP_006571.1

View Full Product Details