Product Details

SNP ID
rs140807328
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:170235314 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCGAGACATGTGTTCTTTTGACAA[C/G]GAACAGCTCTTCACCATGAAATGGA
Phenotype
MIM: 600539
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
PRKCI PubMed Links

Gene Details

Gene
PRKCI
Gene Name
protein kinase C iota
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002740.5 424 Silent Mutation AAC,AAG N62K NP_002731.4
XM_017006860.1 424 UTR 5 XP_016862349.1

View Full Product Details