Product Details
- SNP ID
-
rs140833914
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:47236486 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GACACTGATCCACTAGGTGCTGGCA[A/G]TACTGGATCTCAGCCCTGAGGTCAC
- Phenotype
-
MIM: 607910
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
KIF9
PubMed Links
Gene Details
- Gene
- KIF9
- Gene Name
- kinesin family member 9
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001134878.1 |
2391 |
Silent Mutation |
TAC,TAT |
Y686Y |
NP_001128350.1 |
NM_022342.4 |
2391 |
Silent Mutation |
TAC,TAT |
Y621Y |
NP_071737.1 |
NM_182902.3 |
2391 |
Silent Mutation |
TAC,TAT |
Y686Y |
NP_878905.2 |
XM_005265388.4 |
2391 |
Silent Mutation |
TAC,TAT |
Y719Y |
XP_005265445.1 |
XM_005265389.4 |
2391 |
Silent Mutation |
TAC,TAT |
Y654Y |
XP_005265446.1 |
XM_006713291.3 |
2391 |
Silent Mutation |
TAC,TAT |
Y644Y |
XP_006713354.1 |
XM_011534003.2 |
2391 |
Silent Mutation |
TAC,TAT |
Y700Y |
XP_011532305.1 |
XM_011534004.2 |
2391 |
Silent Mutation |
TAC,TAT |
Y686Y |
XP_011532306.1 |
XM_011534005.2 |
2391 |
Silent Mutation |
TAC,TAT |
Y611Y |
XP_011532307.1 |
XM_011534006.2 |
2391 |
Silent Mutation |
TAC,TAT |
Y593Y |
XP_011532308.1 |
XM_011534007.2 |
2391 |
Silent Mutation |
TAC,TAT |
Y363Y |
XP_011532309.1 |
XM_017007029.1 |
2391 |
Silent Mutation |
TAC,TAT |
Y686Y |
XP_016862518.1 |
XM_017007030.1 |
2391 |
Silent Mutation |
TAC,TAT |
Y611Y |
XP_016862519.1 |
XM_017007031.1 |
2391 |
Silent Mutation |
TAC,TAT |
Y546Y |
XP_016862520.1 |
- Gene
- KIF9-AS1
- Gene Name
- KIF9 antisense RNA 1
There are no transcripts associated with this gene.
View Full Product Details