Product Details

SNP ID
rs141012828
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:184344570 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCCACCGGTTCTCCCGGCCTGTGC[A/G]CCAGGGCTCCGTGGAGCCTGAGAGC
Phenotype
MIM: 600570 MIM: 600495
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CLCN2 PubMed Links

Gene Details

Gene
CLCN2
Gene Name
chloride voltage-gated channel 2
There are no transcripts associated with this gene.

Gene
EIF4G1
Gene Name
eukaryotic translation initiation factor 4 gamma 1
There are no transcripts associated with this gene.

Gene
FAM131A
Gene Name
family with sequence similarity 131 member A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001171093.1 912 Missense Mutation CAC,CGC H149R NP_001164564.1
NM_144635.4 912 Missense Mutation CAC,CGC H234R NP_653236.3
XM_005247113.2 912 Missense Mutation CAC,CGC H149R XP_005247170.1
XM_005247114.3 912 Missense Mutation CAC,CGC H149R XP_005247171.1
XM_011512413.2 912 Missense Mutation CAC,CGC H149R XP_011510715.1

View Full Product Details