Product Details

SNP ID
rs141393714
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:122806907 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCCTTCCTTATGGTTTTGGGAACT[G/T]GTCTAAGATGCATACCTATATCAGA
Phenotype
MIM: 602773
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
DIRC2 PubMed Links

Gene Details

Gene
DIRC2
Gene Name
disrupted in renal carcinoma 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032839.2 533 Missense Mutation GGT,TGT G132C NP_116228.1

View Full Product Details