Product Details

SNP ID
rs141421045
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:48467856 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTCCAATTCCAGCAGCCGAGGCCA[A/G]GAAGAAAGGGGGCAGCAGCTCCAAA
Phenotype
MIM: 606605 MIM: 607290 MIM: 606609
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ATRIP PubMed Links

Gene Details

Gene
ATRIP
Gene Name
ATR interacting protein
There are no transcripts associated with this gene.

Gene
SHISA5
Gene Name
shisa family member 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001272065.1 2290 Intron NP_001258994.1
NM_001272066.1 2290 UTR 3 NP_001258995.1
NM_001272067.1 2290 UTR 3 NP_001258996.1
NM_001272068.1 2290 UTR 3 NP_001258997.1
NM_001272082.1 2290 UTR 3 NP_001259011.1
NM_001272083.1 2290 UTR 3 NP_001259012.1
NM_016479.4 2290 Intron NP_057563.3
XM_005265205.1 2290 Intron XP_005265262.1
XM_006713188.2 2290 Intron XP_006713251.1
XM_011533796.1 2290 Intron XP_011532098.1
Gene
TREX1
Gene Name
three prime repair exonuclease 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007248.3 2290 Intron NP_009179.2
NM_016381.5 2290 Intron NP_057465.1
NM_033629.4 2290 Intron NP_338599.1

View Full Product Details