Product Details

SNP ID
rs141553706
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.3:48862638 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGTCCAAGGTACCAGTGTACTTGC[C/T]TTCTCCTGAAGAAGCCTGAATCTGG
Phenotype
MIM: 613698
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
SLC25A20 PubMed Links

Gene Details

Gene
SLC25A20
Gene Name
solute carrier family 25 member 20
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000387.5 638 Missense Mutation AGC,GGC S147G NP_000378.1

View Full Product Details