Product Details

SNP ID
rs141742243
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:11277314 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTTCACATAAGGCTCTTCATGATA[C/T]GGCCTTTGCCTGCTTTTATGGCATG
Phenotype
MIM: 608760
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ATG7 PubMed Links

Gene Details

Gene
ATG7
Gene Name
autophagy related 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001136031.2 Intron NP_001129503.2
NM_001144912.1 Intron NP_001138384.1
NM_006395.2 Intron NP_006386.1
XM_006712931.3 Intron XP_006712994.1
XM_006712932.3 Intron XP_006712995.1
XM_006712933.3 Intron XP_006712996.1
XM_011533277.2 Intron XP_011531579.1
XM_011533278.2 Intron XP_011531580.1
XM_011533279.2 Intron XP_011531581.1
XM_011533280.2 Intron XP_011531582.1
XM_011533281.2 Intron XP_011531583.1
XM_011533282.2 Intron XP_011531584.1
XM_011533283.2 Intron XP_011531585.1
XM_011533284.2 Intron XP_011531586.1
XM_011533285.2 Intron XP_011531587.1
XM_011533286.2 Intron XP_011531588.1
XM_017005542.1 Intron XP_016861031.1
XM_017005543.1 Intron XP_016861032.1
XM_017005544.1 Intron XP_016861033.1
XM_017005545.1 Intron XP_016861034.1
XM_017005546.1 Intron XP_016861035.1
XM_017005547.1 Intron XP_016861036.1
XM_017005548.1 Intron XP_016861037.1
XM_017005549.1 Intron XP_016861038.1
XM_017005550.1 Intron XP_016861039.1
XM_017005551.1 Intron XP_016861040.1
XM_017005552.1 Intron XP_016861041.1
XM_017005553.1 Intron XP_016861042.1
XM_017005554.1 Intron XP_016861043.1

View Full Product Details