Product Details

SNP ID
rs142637200
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:51993780 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAAGCCTTTGTAGGGGCCTGGGTA[C/T]GTTTGGGAGCCTGAGCTGGAACTGA
Phenotype
MIM: 104620 MIM: 601832
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ABHD14A-ACY1 PubMed Links

Gene Details

Gene
ABHD14A-ACY1
Gene Name
ABHD14A-ACY1 readthrough
There are no transcripts associated with this gene.

Gene
ACY1
Gene Name
aminoacylase 1
There are no transcripts associated with this gene.

Gene
RPL29
Gene Name
ribosomal protein L29
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000992.2 Intron NP_000983.1

View Full Product Details