Product Details

SNP ID
rs142643319
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:62326319 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAAGAATATGTTTAATAAATGCTTA[C/T]ATTGTACCAATGACCCTGTATCTCA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C3orf14 PubMed Links

Gene Details

Gene
C3orf14
Gene Name
chromosome 3 open reading frame 14
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001291941.1 Intron NP_001278870.1
NM_001291942.1 Intron NP_001278871.1
NM_001291943.1 Intron NP_001278872.1
NM_020685.4 Intron NP_065736.1
XM_017006931.1 Intron XP_016862420.1
Gene
PTPRG-AS1
Gene Name
PTPRG antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details