Product Details

SNP ID
rs142938860
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:113651588 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAAAAATTATATCCAAATTAACAC[C/T]ACATACCATATAACAAATTTCTTCC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
USF3 PubMed Links

Gene Details

Gene
USF3
Gene Name
upstream transcription factor family member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001009899.3 10400 UTR 3 NP_001009899.3
XM_005247208.4 10400 UTR 3 XP_005247265.2
XM_017005871.1 10400 UTR 3 XP_016861360.1
XM_017005872.1 10400 UTR 3 XP_016861361.1

View Full Product Details