Product Details

SNP ID
rs143192854
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:88150290 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAATGTTGGCCGCAGCCGCAGATCC[A/G]CAGCTCTGGAACCACTACGCCGACT
Phenotype
MIM: 603363
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C3orf38 PubMed Links

Gene Details

Gene
C3orf38
Gene Name
chromosome 3 open reading frame 38
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_173824.3 Intron NP_776185.2
XM_005264745.4 Intron XP_005264802.1
Gene
CGGBP1
Gene Name
CGG triplet repeat binding protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001008390.1 Intron NP_001008391.1
NM_001195308.1 Intron NP_001182237.1
NM_003663.3 Intron NP_003654.3
XM_017007355.1 Intron XP_016862844.1
Gene
ZNF654
Gene Name
zinc finger protein 654
There are no transcripts associated with this gene.

View Full Product Details