Product Details

SNP ID
rs143323039
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:126989166 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAAGCTCTTCGTGGGCACACCCAT[C/T]GATGGCAAGTCCGAGTACTTCCCCA
Phenotype
MIM: 601055
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PLXNA1 PubMed Links

Gene Details

Gene
PLXNA1
Gene Name
plexin A1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032242.3 1268 Silent Mutation ATC,ATT I191I NP_115618.3
XM_011512908.2 1268 Silent Mutation ATC,ATT I209I XP_011511210.1
XM_011512909.1 1268 Silent Mutation ATC,ATT I209I XP_011511211.1

View Full Product Details