Product Details

SNP ID
rs143517124
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:32526908 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGAACATCTAAGACAGGCTTCTGG[A/C]CTACATTGAAGAAAAAGAAAAAAAA
Phenotype
MIM: 615890
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
DYNC1LI1 PubMed Links

Gene Details

Gene
DYNC1LI1
Gene Name
dynein cytoplasmic 1 light intermediate chain 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016141.3 1567 Missense Mutation GGC,GTC G488V NP_057225.2

View Full Product Details