Product Details

SNP ID
rs144206255
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:164982291 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGGAAGCGAATTTTTATTTCCGTT[A/G]TACGTTAGAGTAACTGCATTGACAG
Phenotype
MIM: 609845
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SI PubMed Links

Gene Details

Gene
SI
Gene Name
sucrase-isomaltase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001041.3 5434 Silent Mutation TAC,TAT Y1789Y NP_001032.2
XM_011513078.2 5434 Silent Mutation TAC,TAT Y1756Y XP_011511380.1

View Full Product Details