Product Details

SNP ID
rs144623757
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:132562734 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCATCTCTATACCTCTTTGGTGAT[A/G]TGTCTGTTAAAGTCTTTTGTCCATT
Phenotype
MIM: 614288
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ACAD11 PubMed Links

Gene Details

Gene
ACAD11
Gene Name
acyl-CoA dehydrogenase family member 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032169.4 Intron NP_115545.3
Gene
NPHP3-ACAD11
Gene Name
NPHP3-ACAD11 readthrough (NMD candidate)
There are no transcripts associated with this gene.

View Full Product Details