Product Details

SNP ID
rs144737998
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:94062352 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTCCTAATCACTGTTGTAACCGCT[C/T]TACATGCCTACTTCAGTTTAACACT
Phenotype
MIM: 608922 MIM: 616588
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ARL13B PubMed Links

Gene Details

Gene
ARL13B
Gene Name
ADP ribosylation factor like GTPase 13B
There are no transcripts associated with this gene.

Gene
DHFR2
Gene Name
dihydrofolate reductase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001195643.1 Intron NP_001182572.1
NM_176815.4 Intron NP_789785.1
XM_011512537.2 Intron XP_011510839.1
Gene
NSUN3
Gene Name
NOP2/Sun RNA methyltransferase family member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022072.3 Intron NP_071355.1

View Full Product Details