Product Details

SNP ID
rs144747067
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:126572259 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTCTTGACCTTTCCCCTCTGGCTG[C/T]CCAGGCTCTCCAGAAAGACATTTTG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TXNRD3NB PubMed Links

Gene Details

Gene
TXNRD3NB
Gene Name
thioredoxin reductase 3 neighbor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001039783.1 764 Silent Mutation GGA,GGG G95G NP_001034872.1

View Full Product Details