Product Details

SNP ID
rs144792046
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:156540193 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTTATTAAGCAAGACTTTTCACAA[A/G]AATTGAGTATAAATTCAATCTCCAT
Phenotype
MIM: 601141 MIM: 606213
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KCNAB1 PubMed Links

Gene Details

Gene
KCNAB1
Gene Name
potassium voltage-gated channel subfamily A member regulatory beta subunit 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001308217.1 5776 Intron NP_001295146.1
NM_001308222.1 5776 Intron NP_001295151.1
NM_003471.3 5776 Intron NP_003462.2
NM_172159.3 5776 Intron NP_751891.1
NM_172160.2 5776 Intron NP_751892.1
XM_011513115.2 5776 UTR 3 XP_011511417.1
XM_011513116.2 5776 Intron XP_011511418.1
XM_011513117.2 5776 UTR 3 XP_011511419.1
XM_017007171.1 5776 UTR 3 XP_016862660.1
XM_017007172.1 5776 UTR 3 XP_016862661.1
XM_017007173.1 5776 UTR 3 XP_016862662.1
XM_017007174.1 5776 Intron XP_016862663.1
Gene
SSR3
Gene Name
signal sequence receptor subunit 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001308197.1 5776 UTR 3 NP_001295126.1
NM_001308204.1 5776 UTR 3 NP_001295133.1
NM_001308205.1 5776 UTR 3 NP_001295134.1
NM_007107.4 5776 UTR 3 NP_009038.1

View Full Product Details