Product Details

SNP ID
rs144938011
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:185491628 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTAAATTTTTTAATGAGGGTTCCA[A/G]GAATTAATGCCACCATGGCAATGGC
Phenotype
MIM: 604915
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MAP3K13 PubMed Links

Gene Details

Gene
MAP3K13
Gene Name
mitogen-activated protein kinase kinase kinase 13
There are no transcripts associated with this gene.

Gene
TMEM41A
Gene Name
transmembrane protein 41A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_080652.3 638 Missense Mutation CCT,CTT P235L NP_542383.1
XM_017007437.1 638 Missense Mutation CCT,CTT P181L XP_016862926.1

View Full Product Details