Product Details

SNP ID
rs145121359
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:142308431 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATAGTTCTTCATTCCTCCACCAATC[A/G]GGAAAAAAACCCTTCAATCTTTATT
Phenotype
MIM: 607994
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
XRN1 PubMed Links

Gene Details

Gene
XRN1
Gene Name
5'-3' exoribonuclease 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282857.1 7953 UTR 3 NP_001269786.1
NM_001282859.1 7953 Intron NP_001269788.1
NM_019001.4 7953 UTR 3 NP_061874.3
XM_006713673.2 7953 Intron XP_006713736.1
XM_011512919.2 7953 UTR 3 XP_011511221.1
XM_011512920.2 7953 UTR 3 XP_011511222.1
XM_011512922.2 7953 UTR 3 XP_011511224.1
XM_017006640.1 7953 UTR 3 XP_016862129.1
XM_017006641.1 7953 UTR 3 XP_016862130.1
XM_017006642.1 7953 Intron XP_016862131.1

View Full Product Details