Product Details

SNP ID
rs145282095
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:98283002 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCCCTGTTCTTGGTGTTCTTGGTG[A/C]TCTATCTCATCACTATTGTGTGGAA
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
OR5H2 PubMed Links

Gene Details

Gene
OR5H2
Gene Name
olfactory receptor family 5 subfamily H member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005482.1 115 Missense Mutation ATC,CTC I39L NP_001005482.1

View Full Product Details