Product Details
- SNP ID
-
rs145475632
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:190388196 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGCAACTCAAGACACCTGCAGCAGG[A/G]CGTGAGAAAAAGTAAAAGACCAGTA
- Phenotype
-
MIM: 603959
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CLDN16
PubMed Links
Gene Details
- Gene
- CLDN16
- Gene Name
- claudin 16
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_006580.3 |
325 |
Missense Mutation |
GAC,GGC |
D26G |
NP_006571.1 |
View Full Product Details