Product Details

SNP ID
rs145795547
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:58428510 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGCACAGATTTCAGCTCCTACTCC[A/G]AACTGTGGCCAGCCTCCTTCCACAG
Phenotype
MIM: 179060 MIM: 611450
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PDHB PubMed Links

Gene Details

Gene
PDHB
Gene Name
pyruvate dehydrogenase (lipoamide) beta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000925.3 886 Silent Mutation TTC,TTT F299F NP_000916.2
NM_001173468.1 886 Silent Mutation TTC,TTT F281F NP_001166939.1
NM_001315536.1 886 Silent Mutation TTC,TTT F281F NP_001302465.1
Gene
PXK
Gene Name
PX domain containing serine/threonine kinase like
There are no transcripts associated with this gene.

View Full Product Details