Product Details

SNP ID
rs145922036
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:51978310 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGCCTTTAACTCTCACACGTGGGA[G/T]CAGCTGGGCACACTGCAGCTACTGT
Phenotype
MIM: 104620
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ABHD14A PubMed Links

Gene Details

Gene
ABHD14A
Gene Name
abhydrolase domain containing 14A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015407.4 443 Missense Mutation GAG,GAT E111D NP_056222.2
Gene
ABHD14A-ACY1
Gene Name
ABHD14A-ACY1 readthrough
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001316331.1 443 Missense Mutation AGC,ATC S63I NP_001303260.1
Gene
ABHD14B
Gene Name
abhydrolase domain containing 14B
There are no transcripts associated with this gene.

Gene
ACY1
Gene Name
aminoacylase 1
There are no transcripts associated with this gene.

View Full Product Details