Product Details

SNP ID
rs146036883
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:112991264 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCGGACAACATCTTCACGGCCACT[C/G]AACGGAACCGCATCGACTACGTCAG
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
GTPBP8 PubMed Links

Gene Details

Gene
GTPBP8
Gene Name
GTP binding protein 8 (putative)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014170.2 312 Missense Mutation CAA,GAA Q89E NP_054889.2
NM_138485.1 312 Missense Mutation CAA,GAA Q89E NP_612494.1

View Full Product Details