Product Details

SNP ID
rs146112611
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:33114245 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCGGCACGCGCTGGACAAGTACAG[C/G]GGCGAGCACTGGGCCGAGAGCGTGG
Phenotype
MIM: 605497
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CRTAP PubMed Links

Gene Details

Gene
CRTAP
Gene Name
cartilage associated protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006371.4 288 Silent Mutation AGC,AGG S56R NP_006362.1

View Full Product Details