Product Details

SNP ID
rs146340094
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:49022706 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGTGCCTCCTCCCTGCACAGGTGG[G/T]ATCCCACCAGGACATCACCGAAGAC
Phenotype
MIM: 146691 MIM: 615150 MIM: 612911 MIM: 606031
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
DALRD3 PubMed Links

Gene Details

Gene
DALRD3
Gene Name
DALR anticodon binding domain containing 3
There are no transcripts associated with this gene.

Gene
IMPDH2
Gene Name
inosine monophosphate dehydrogenase 2
There are no transcripts associated with this gene.

Gene
MIR191
Gene Name
microRNA 191
There are no transcripts associated with this gene.

Gene
MIR425
Gene Name
microRNA 425
There are no transcripts associated with this gene.

Gene
NDUFAF3
Gene Name
NADH:ubiquinone oxidoreductase complex assembly factor 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_199069.1 294 Missense Mutation GGA,GTA G92V NP_951032.1
NM_199070.1 294 Missense Mutation GGA,GTA G35V NP_951033.1
NM_199073.1 294 Missense Mutation GGA,GTA G35V NP_951047.1
NM_199074.1 294 Missense Mutation GGA,GTA G35V NP_951056.1
Gene
QRICH1
Gene Name
glutamine rich 1
There are no transcripts associated with this gene.

Gene
WDR6
Gene Name
WD repeat domain 6
There are no transcripts associated with this gene.

View Full Product Details